Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133683.3 | NP_598444.1 | 4 | 509 | ||
GenBank mRNA | AK018383.1 | - | 4 | 458 | ||
AK034298.1 | - | 4 | 515 | |||
AK036228.1 | - | 3 | 586 | |||
AK042426.1 | - | 3 | 564 | |||
AK079735.1 | - | 367 | ||||
AK136778.1 | - | 498 | ||||
AK138931.1 | - | 4 | 497 | |||
AK155275.1 | - | 4 | 470 | |||
AK170143.1 | - | 4 | 469 | |||
AK171087.1 | - | 4 | 469 | |||
AK209903.1 | - | 3 | 255 | |||
AK213890.1 | - | 3 | 255 | |||
BC016895.1 | - | 4 | 511 | 4239007 |