Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam220a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026050.2 | NP_080326.2 | 3 | 988 | ||
NM_133703.4 | NP_598464.3 | 2 | 855 | |||
GenBank mRNA | AB374429.1 | - | 2 | 629 | ||
AK014805.1 | - | 3 | 921 | |||
AK075875.1 | - | 2 | 841 | |||
AK076127.1 | - | 2 | 838 | |||
AK156614.1 | - | 3 | 933 | |||
AY714985.1 | - | 1 | 404 | |||
BC005638.1 | - | 1 | 117 | |||
BC085297.1 | - | 2 | 745 | |||
BC100440.1 | - | 2 | 800 | |||
BC147186.1 | - | 2 | 457 | 9056217 | ||
BC147187.1 | - | 2 | 457 | 9056219 | ||
BC172003.1 | - | 2 | 457 | 9054289 |