Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042556.1 | NP_001036021.1 | 8 | 998 | ||
NM_023323.3 | NP_075812.3 | 8 | 648 | |||
GenBank mRNA | AB041810.1 | - | 8 | 1028 | ||
AK010776.1 | - | 8 | 642 | |||
AK011644.1 | - | 8 | 626 | |||
AK012188.1 | - | 8 | 648 | |||
AK012641.1 | - | 8 | 627 | |||
AK013395.1 | - | 8 | 623 | |||
AK017678.1 | - | 8 | 619 | |||
AK051135.1 | - | 8 | 999 |