Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Marc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133684.3 | NP_598445.1 | 5, 6 | 856 | ||
GenBank mRNA | AK002874.1 | - | 5, 6 | 853 | ||
AK013440.1 | - | 5, 6 | 876 | |||
AK017150.1 | - | 5, 6 | 856 | |||
AK051230.1 | - | 5, 6 | 857 | |||
AK184682.1 | - | 2, 3 | 202 |