Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026056.4 | NP_080332.1 | 3 | 460 | ||
GenBank mRNA | AK010235.1 | - | 3 | 207 | ||
AK013331.1 | - | 3 | 217 | |||
AK033071.1 | - | 3 | 216 | |||
AK035556.1 | - | 3 | 461 | |||
AK047701.1 | - | 1 | 27 | |||
AK049813.1 | - | 3 | 239 | |||
BC050752.1 | - | 3 | 221 | 6533812 | ||
BC057937.1 | - | 3 | 275 | 5325599 |