Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026065.3 | NP_080341.1 | 5 | 436 | ||
GenBank mRNA | AK002530.1 | - | 5 | 421 | ||
AK003427.2 | - | 5 | 437 | |||
AK013693.1 | - | 5 | 433 | |||
BC024337.1 | - | 5 | 417 | 4984291 |