Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026069.3 | NP_080345.1 | 3 | 234 | ||
GenBank mRNA | AK002592.1 | - | 3 | 203 | ||
AK008563.1 | - | 3 | 203 | |||
AK011101.1 | - | 3 | 195 | |||
AK012544.1 | - | 3 | 203 | |||
AK014003.1 | - | 3 | 203 | |||
AK019020.1 | - | 3 | 203 | |||
AK019294.1 | - | 3 | 201 | |||
AK028138.1 | - | 3 | 193 | |||
AK168550.1 | - | 3 | 203 | |||
BC034524.1 | - | 181 | ||||
BC054388.1 | - | 3 | 190 | 6478626 | ||
BC081438.1 | - | 2 | 165 | 6817584 |
Gene Symbol: Rpl37rt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271590.1 | NP_001258519.1 | 2 | 248 | ||
NM_001271591.1 | NP_001258520.1 | 2 | 242 | |||
GenBank mRNA | CO799256.1 | - | 280 | |||
CO814488.2 | - | 1 | 134 |