Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005502.3 | NP_005493.2 | 35 | 5120 | ||
XM_005251773.2 | XP_005251830.1 | 35 | 5035 | |||
XM_005251776.2 | XP_005251833.1 | 34 | 4875 | |||
XM_011518339.2 | XP_011516641.1 | 36 | 5110 | |||
XM_011518340.2 | XP_011516642.1 | 37 | 5325 | |||
XM_011518341.2 | XP_011516643.1 | 36 | 5104 | |||
XM_011518342.2 | XP_011516644.1 | 33 | 4813 | |||
XM_017014378.1 | XP_016869867.1 | 36 | 5308 | |||
XM_017014379.1 | XP_016869868.1 | 36 | 5716 | |||
XM_017014380.1 | XP_016869869.1 | 36 | 5042 | |||
XM_017014381.1 | XP_016869870.1 | 36 | 5108 | |||
XM_017014382.1 | XP_016869871.1 | 34 | 5573 | |||
GenBank mRNA | AB055982.1 | - | 34 | 4716 | ||
AB445477.1 | - | 34 | 4716 | |||
AF165281.1 | - | 33 | 4656 | |||
AF285167.1 | - | 35 | 5006 | |||
AJ012376.1 | - | 33 | 4656 | |||
BC146856.1 | - | 35 | 4930 | 9056784 |