Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205239.1 | NP_001192168.1 | 10 | 1186 | ||
NM_029922.3 | NP_084198.2 | 10 | 1186 | |||
GenBank mRNA | AK035349.1 | - | 10 | 1110 | ||
AK076321.1 | - | 10 | 1110 | |||
BC031387.1 | - | 9 | 857 | 5038718 | ||
BC052064.1 | - | 10 | 1184 | |||
BC058266.1 | - | 1 | 80 | |||
BC062096.1 | - | 9 | 1043 | 5716648 |