Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023852.5 | NP_076341.1 | 2 | 387 | ||
GenBank mRNA | AB232589.1 | - | 2 | 199 | ||
AF312037.1 | - | 2 | 199 | |||
AJ310532.1 | - | 2 | 199 | |||
AK012468.1 | - | 2 | 380 | |||
AK014050.1 | - | 2 | 366 | |||
AK014132.1 | - | 2 | 309 | |||
AK079574.1 | - | 2 | 284 | |||
AK148904.1 | - | 2 | 308 | |||
AK161006.1 | - | 2 | 285 | |||
AK164316.1 | - | 2 | 332 | |||
AY026947.1 | - | 2 | 268 | |||
BC114335.2 | - | 2 | 260 | 40045744 |