Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gprasp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004359.2 | NP_001004359.1 | 10 | 3165 | ||
NM_001005385.1 | NP_001005385.1 | 10 | 3051 | |||
NM_026081.5 | NP_080357.4 | 11 | 3254 | |||
GenBank mRNA | AB093241.1 | - | 5 | 2371 | ||
BC027187.1 | - | 1 | 493 | 4505972 | ||
BC058970.1 | - | 1 | 2008 | 6826050 | ||
BC070413.1 | - | 11 | 3202 | |||
BC085157.1 | - | 10 | 2953 | 30357108 |