Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dock7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290636.1 | NP_001277565.1 | 25 | 3126 | ||
NM_026082.5 | NP_080358.3 | 24 | 3036 | |||
GenBank mRNA | BC086672.1 | - | 5 | 544 | 30647593 | |
BC094420.1 | - | 12 | 1322 | |||
CB246838.1 | - | 4 | 292 | |||
CD802819.1 | - | 5 | 688 |