Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc2hc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173181.3 | NP_775273.1 | 5 | 547 | ||
GenBank mRNA | AK014202.1 | - | 5 | 483 | ||
AK083941.1 | - | 5 | 491 | |||
AK085187.1 | - | 5 | 485 | |||
AK170609.1 | - | 5 | 488 | |||
BC044809.1 | - | 5 | 469 | |||
BC087731.1 | - | 511 | 6850296 |