Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbld2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026085.2 | NP_080361.1 | 10 | 1081 | ||
GenBank mRNA | AK007784.1 | - | 9 | 836 | ||
AK014198.1 | - | 10 | 1082 | |||
AK018692.1 | - | 9 | 837 | |||
BC061073.1 | - | 9 | 836 | 30381600 |