Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyzl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026092.3 | NP_080368.1 | 3, 4 | 442 | ||
GenBank mRNA | AK006241.1 | - | 3, 4 | 443 | ||
AK006630.1 | - | 3, 4 | 443 | |||
AY572447.1 | - | 3, 4 | 401 | |||
BC048450.1 | - | 3, 4 | 413 | 6774504 |