Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hypm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026096.1 | NP_080372.1 | 1 | 201 | ||
GenBank mRNA | AK006319.1 | - | 1 | 190 | ||
AK006788.1 | - | 1 | 201 | |||
BC049569.1 | - | 1 | 159 | 6772969 | ||
BC059721.1 | - | 1 | 176 | 6742451 |