Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tctex1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163767.1 | NP_001157239.1 | 3, 4 | 419 | ||
NM_001163768.1 | NP_001157240.1 | 2, 3 | 326 | |||
NM_026100.3 | NP_080376.1 | 3, 4 | 451 | |||
GenBank mRNA | AK006804.1 | - | 3, 4 | 420 | ||
AK015315.1 | - | 3, 4 | 454 | |||
BB634690.1 | - | 2, 3 | 338 | |||
BC048635.1 | - | 3, 4 | 391 | |||
BC099549.1 | - | 3, 4 | 425 | 30919464 | ||
BC132398.1 | - | 3, 4 | 369 | 40130675 | ||
BC132400.1 | - | 3, 4 | 369 | 40130677 | ||
BC145187.1 | - | 3, 4 | 369 | 9053738 |