Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdhd1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026108.3 | NP_080384.2 | 1 | 371 | ||
GenBank mRNA | AK007231.1 | - | 1 | 397 | ||
AK014922.1 | - | 1 | 348 | |||
BC048447.1 | - | 1 | 360 | 6705877 | ||
CN834894.1 | - | 374 |