Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paxbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026110.2 | NP_080386.3 | 10, 11 | 1762 | ||
GenBank mRNA | AK007365.1 | - | 4, 5 | 467 | ||
AK079394.1 | - | 3, 4 | 627 | |||
AY033907.1 | - | 10, 11 | 1547 | |||
AY033908.1 | - | 10, 11 | 1564 | |||
BC014838.1 | - | 3, 4 | 247 | 3588322 | ||
BC027145.1 | - | 2, 3 | 182 |