Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAOB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000898.4 | NP_000889.3 | 3 | 368 | ||
XM_005272608.3 | XP_005272665.1 | 5 | 468 | |||
XM_017029523.1 | XP_016885012.1 | 5 | 518 | |||
XM_017029524.1 | XP_016885013.1 | 4 | 328 | |||
GenBank mRNA | AK294306.1 | - | 4 | 286 | ||
AK298942.1 | - | 3 | 305 | |||
AK310458.1 | - | 3 | 340 | |||
AK312679.1 | - | 3 | 343 | |||
AU127841.1 | - | 3 | 341 | |||
BC022494.1 | - | 3 | 314 | 4800208 | ||
DC326844.1 | - | 3 | 369 | |||
M69177.1 | - | 3 | 269 | |||
S62734.1 | - | 3 | 192 |