Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bag4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026121.3 | NP_080397.1 | 3 | 546 | ||
GenBank mRNA | AF332863.1 | - | 3 | 539 | ||
AK010765.1 | - | 3 | 537 | |||
AK136899.1 | - | 3 | 536 | |||
AK160316.1 | - | 3 | 538 | |||
AK162658.1 | - | 3 | 533 | |||
BC037239.1 | - | 3 | 544 | |||
BC058518.1 | - | 3 | 529 | 5717401 | ||
BC138201.1 | - | 3 | 624 | 8861220 | ||
BC145986.1 | - | 3 | 624 | 40131289 |