Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdlim7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114087.2 | NP_001107559.1 | 6, 7 | 620 | ||
NM_001114088.2 | NP_001107560.1 | 6, 7 | 620 | |||
NM_026131.4 | NP_080407.3 | 6, 7 | 518 | |||
GenBank mRNA | AK003019.1 | - | 7, 8 | 594 | ||
AK003338.1 | - | 6, 7 | 509 | |||
AK010141.1 | - | 6, 7 | 507 | |||
AK033091.1 | - | 612 | ||||
AK077998.1 | - | 6, 7 | 612 | |||
AK128987.1 | - | 6, 7 | 608 | |||
AK142933.1 | - | 6, 7 | 604 | |||
AK167476.1 | - | 6, 7 | 612 | |||
BC049565.1 | - | 6, 7 | 506 | 6588160 | ||
BC052698.1 | - | 6, 7 | 591 |