Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930449I24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026136.2 | NP_080412.1 | 790 | |||
GenBank mRNA | AK015434.1 | - | 791 | |||
BC147588.1 | - | 787 | 9056578 | |||
BC147604.1 | - | 787 | 9056594 |
Gene Symbol: Gm6370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243110.1 | NP_001230039.1 | 5 | 777 |