Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024200.4 | NP_077162.2 | 12 | 1485 | ||
GenBank mRNA | AK018181.1 | - | 12 | 1482 | ||
AK081358.1 | - | 5 | 471 | |||
AK141611.1 | - | 12 | 1486 | |||
AK220169.1 | - | 12 | 1473 | |||
AY174062.1 | - | 11 | 1614 | |||
BC008137.1 | - | 8 | 752 | |||
BC047050.1 | - | 3 | 220 | |||
BC056641.1 | - | 12 | 1444 | 5097345 |