Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024200.4 | NP_077162.2 | 8 | 1055 | ||
GenBank mRNA | AK018181.1 | - | 8 | 1052 | ||
AK081358.1 | - | 1 | 41 | |||
AK141611.1 | - | 8 | 1056 | |||
AK220169.1 | - | 8 | 1043 | |||
AY174062.1 | - | 7 | 1184 | |||
BC008137.1 | - | 4 | 322 | |||
BC056641.1 | - | 8 | 1014 | 5097345 |