Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024200.4 | NP_077162.2 | 3 | 359 | ||
GenBank mRNA | AK018181.1 | - | 3 | 356 | ||
AK046313.1 | - | 3 | 312 | |||
AK141611.1 | - | 3 | 360 | |||
AK220169.1 | - | 3 | 347 | |||
AY174062.1 | - | 2 | 488 | |||
BC056641.1 | - | 3 | 318 | 5097345 |