Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ears2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014324.2 | - | 174 | |||
AK076043.1 | - | 2 | 174 | |||
AK129469.1 | - | 140 | ||||
AK167506.1 | - | 2 | 186 | |||
AK169225.1 | - | 2 | 175 | |||
BC025907.1 | - | 2 | 192 | 5064949 | ||
BY122579.1 | - | 2 | 215 |