Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ears2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014324.2 | - | 859 | |||
AK129469.1 | - | 825 | ||||
AK167506.1 | - | 4 | 871 | |||
AK169225.1 | - | 4 | 860 | |||
BC025907.1 | - | 4 | 877 | 5064949 |