Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppil4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026141.3 | NP_080417.2 | 5 | 419 | ||
GenBank mRNA | AK011443.1 | - | 5 | 391 | ||
AK012371.1 | - | 5 | 396 | |||
AK013377.1 | - | 5 | 392 | |||
AK014406.2 | - | 392 | ||||
AK014453.1 | - | 4 | 944 | |||
AK017370.1 | - | 5 | 419 | |||
AK167168.1 | - | 5 | 384 | |||
AK167258.1 | - | 5 | 391 | |||
BC079912.1 | - | 5 | 381 | 30661438 |