Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhdds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026144.4 | NP_080420.2 | 8 | 883 | ||
GenBank mRNA | AK004993.1 | - | 8 | 943 | ||
AK014317.1 | - | 9 | 1071 | |||
AK017556.1 | - | 8 | 955 | |||
AK034277.1 | - | 8 | 945 | |||
AK036929.1 | - | 8 | 961 | |||
AK049991.1 | - | 8 | 884 | |||
AK084682.1 | - | 1 | 459 | |||
AK135087.1 | - | 8 | 945 | |||
AK149138.1 | - | 8 | 915 | |||
AK171207.1 | - | 8 | 957 | |||
BC004011.1 | - | 8 | 931 | 3493585 |