Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026155.3 | NP_080431.1 | 3 | 314 | ||
GenBank mRNA | AK002818.1 | - | 3 | 315 | ||
AK029353.1 | - | 3 | 315 | |||
AK076235.1 | - | 3 | 347 | |||
AK088555.1 | - | 3 | 319 | |||
AK150255.1 | - | 3 | 308 | |||
AK162235.1 | - | 1 | 18 | |||
AK192841.1 | - | 3 | 235 | |||
AK194871.1 | - | 3 | 235 | |||
AK206034.1 | - | 3 | 235 | |||
BC011111.1 | - | 3 | 321 | 3965897 | ||
BC020335.1 | - | 3 | 286 | |||
CT010172.1 | - | 3 | 298 |