Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175118.3 | NP_780327.1 | 2 | 1018 | ||
GenBank mRNA | AK002934.1 | - | 2 | 1017 | ||
AK088940.1 | - | 2 | 962 | |||
AK163560.1 | - | 2 | 1027 | |||
AW493803.1 | - | 1 | 26 |