Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026163.2 | NP_080439.1 | 11 | 2092 | ||
GenBank mRNA | AK004650.1 | - | 11 | 2093 | ||
AK005020.1 | - | 11 | 2124 | |||
AK049481.1 | - | 11 | 2093 | |||
AK167660.1 | - | 11 | 2094 | |||
BC018487.1 | - | 5 | 648 | |||
BC037133.1 | - | 6 | 698 | |||
BC068279.1 | - | 9 | 1373 | |||
BC138252.1 | - | 11 | 2092 | 8861271 | ||
BC138253.1 | - | 11 | 2092 | 8861272 |