Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nvl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026171.1 | NP_080447.1 | 14 | 1704 | ||
GenBank mRNA | AK004676.1 | - | 14 | 1736 | ||
AK054502.1 | - | 9 | 1183 | |||
AK140509.1 | - | 14 | 1735 | |||
BC031847.1 | - | 14 | 1687 | 3964470 |