Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026173.3 | NP_080449.1 | 3 | 438 | ||
GenBank mRNA | AK004769.2 | - | 3 | 366 | ||
AK012926.1 | - | 3 | 406 | |||
AK089013.1 | - | 3 | 374 | |||
AK098149.1 | - | 3 | 331 | |||
AK150656.1 | - | 3 | 439 | |||
BC016527.1 | - | 3 | 352 | 3157281 | ||
BC027118.1 | - | 3 | 376 | 5359153 | ||
BC037631.1 | - | 3 | 395 | 4954391 |