Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026174.3 | NP_080450.1 | 10 | 1350 | ||
GenBank mRNA | AB093238.1 | - | 10 | 1268 | ||
AK004761.1 | - | 10 | 1317 | |||
AK031065.1 | - | 5 | 493 | |||
AK045342.1 | - | 1329 | ||||
BC006924.1 | - | 8 | 1075 | |||
BC026420.1 | - | 6 | 617 | |||
BC043134.1 | - | 10 | 1302 | 6811715 |