Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026174.3 | NP_080450.1 | 3 | 318 | ||
GenBank mRNA | AB093238.1 | - | 3 | 260 | ||
AK004761.1 | - | 3 | 285 | |||
AK042053.1 | - | 2 | 1759 | |||
AK045342.1 | - | 296 | ||||
AK192189.1 | - | 2 | 191 | |||
AK193291.1 | - | 2 | 190 | |||
AK197306.1 | - | 2 | 190 | |||
AK197494.1 | - | 2 | 191 | |||
AK198046.1 | - | 2 | 190 | |||
AK201574.1 | - | 2 | 194 | |||
AK202043.1 | - | 2 | 190 | |||
AK204612.1 | - | 2 | 190 | |||
AK207715.1 | - | 2 | 211 | |||
AK213995.1 | - | 2 | 190 | |||
BC006924.1 | - | 1 | 67 | |||
BC043134.1 | - | 3 | 270 | 6811715 | ||
BC052090.1 | - | 3 | 271 | |||
BY276189.1 | - | 3 | 320 |