Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026174.3 | NP_080450.1 | 13 | 1937 | ||
GenBank mRNA | AB093238.1 | - | 11 | 4128 | ||
AK004761.1 | - | 13 | 1904 | |||
AK009433.1 | - | 2 | 187 | |||
AK031065.1 | - | 8 | 1080 | |||
AK045342.1 | - | 1916 | ||||
AK169913.1 | - | 2 | 2004 | |||
BC006924.1 | - | 11 | 1662 | |||
BC026420.1 | - | 9 | 1204 | |||
BC043134.1 | - | 13 | 1889 | 6811715 |