Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sf3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026175.5 | NP_080451.4 | 12 | 2010 | ||
GenBank mRNA | AK004763.1 | - | 12 | 1993 | ||
AK008075.1 | - | 1 | 37 | |||
AK028829.1 | - | 12 | 1962 | |||
AK029095.1 | - | 12 | 1980 | |||
AK030115.1 | - | 4 | 536 | |||
AK030223.1 | - | 12 | 1997 | |||
AK134178.1 | - | 12 | 1988 | |||
AK160061.1 | - | 12 | 1993 | |||
BC010727.1 | - | 3 | 465 | |||
BC029753.1 | - | 12 | 1974 | 2651773 |