Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026178.2 | NP_080454.1 | 5, 6 | 688 | ||
GenBank mRNA | AK004819.1 | - | 5, 6 | 642 | ||
AK011418.1 | - | 5, 6 | 653 | |||
AK033889.1 | - | 5, 6 | 708 | |||
AK149704.1 | - | 5, 6 | 638 | |||
AK158577.1 | - | 5, 6 | 646 | |||
AK171637.1 | - | 5, 6 | 642 | |||
AK193751.1 | - | 3, 4 | 333 | |||
AY424300.1 | - | 5, 6 | 528 | |||
BC021914.1 | - | 5, 6 | 688 | 4506710 | ||
BC034117.1 | - | 6, 7 | 1040 |