Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026179.2 | NP_080455.1 | 5 | 916 | ||
GenBank mRNA | AK004873.1 | - | 5 | 802 | ||
AK007421.1 | - | 5 | 798 | |||
AK019488.2 | - | 3 | 342 | |||
AK028453.1 | - | 5 | 802 | |||
AK050377.1 | - | 5 | 801 | |||
AK152717.1 | - | 6 | 850 | |||
BC006683.1 | - | 2 | 142 | |||
BC037063.1 | - | 5 | 798 | 4985992 |