Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpatch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026181.1 | NP_080457.1 | 5 | 501 | ||
GenBank mRNA | AK004868.1 | - | 5 | 501 | ||
AK014367.1 | - | 5 | 480 | |||
AK154571.1 | - | 5 | 482 | |||
BC100577.1 | - | 5 | 476 | |||
BC125584.1 | - | 5 | 492 | 40130099 | ||
BC132155.1 | - | 5 | 492 | 40130432 |