Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtfr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253390.1 | NP_001240319.1 | 3 | 242 | ||
NM_001253391.1 | NP_001240320.1 | 3 | 404 | |||
NM_026182.5 | NP_080458.1 | 3 | 252 | |||
GenBank mRNA | AF354708.1 | - | 2 | 203 | ||
AK004846.1 | - | 3 | 230 | |||
AK006952.1 | - | 3 | 404 | |||
AK009195.2 | - | 3 | 218 | |||
AK016316.1 | - | 3 | 390 | |||
AK018048.1 | - | 3 | 234 | |||
AK075822.1 | - | 3 | 228 | |||
AK075992.1 | - | 3 | 252 | |||
AK161251.1 | - | 3 | 225 | |||
BC019971.1 | - | 3 | 200 | 4159093 | ||
BC042471.1 | - | 2 | 176 | 3988195 | ||
BY143181.1 | - | 3 | 256 | |||
CK625955.1 | - | 2 | 195 | |||
DQ369744.1 | - | 3 | 363 |