Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cwc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026186.4 | NP_080462.2 | 8 | 1099 | ||
GenBank mRNA | AK004983.2 | - | 8 | 1043 | ||
AK017849.1 | - | 8 | 1039 | |||
BC021478.1 | - | 8 | 1013 | |||
BC053447.1 | - | 1 | 253 | |||
BC079553.1 | - | 8 | 1021 | 6856335 |