Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cwc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026186.4 | NP_080462.2 | 3 | 378 | ||
GenBank mRNA | AK004983.2 | - | 3 | 322 | ||
AK017849.1 | - | 3 | 323 | |||
AK030628.1 | - | 3 | 307 | |||
AK035158.1 | - | 3 | 307 | |||
BC021478.1 | - | 3 | 292 | |||
BC079553.1 | - | 3 | 300 | 6856335 |