Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calcoco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026192.3 | NP_080468.1 | 9 | 1288 | ||
GenBank mRNA | AK007393.1 | - | 9 | 1179 | ||
AK018734.2 | - | 9 | 1174 | |||
AK035598.1 | - | 9 | 1200 | |||
AK087513.1 | - | 9 | 1242 | |||
AK220400.1 | - | 9 | 1201 | |||
AY131329.1 | - | 8 | 1154 | |||
BC054783.1 | - | 9 | 1182 | 6815943 |