Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ufl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026194.4 | NP_080470.2 | 19 | 2335 | ||
GenBank mRNA | AK007993.2 | - | 19 | 2280 | ||
AK013382.1 | - | 12 | 1597 | |||
AK032663.1 | - | 19 | 2271 | |||
AK082785.1 | - | 9 | 1042 | |||
AK129211.1 | - | 18 | 2248 | |||
AK132697.1 | - | 19 | 2299 | |||
BC058809.1 | - | 18 | 3659 | |||
BC099529.1 | - | 19 | 2322 | |||
BC132195.1 | - | 19 | 2287 | 40130472 | ||
BC138153.1 | - | 19 | 2287 | 8861172 |