Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004990.3 | NP_004981.2 | 14 | 1854 | ||
XM_006719398.3 | XP_006719461.1 | 13 | 1689 | |||
GenBank mRNA | AK122956.1 | - | 15 | 2335 | ||
AK222767.1 | - | 14 | 1736 | |||
AK290857.1 | - | 14 | 1725 | |||
AK293945.1 | - | 13 | 1665 | |||
BC002384.2 | - | 14 | 1722 | 2961615 | ||
BC006328.2 | - | 14 | 1722 | 4139330 | ||
BC011639.2 | - | 14 | 1724 | 4120680 | ||
BC011849.2 | - | 14 | 1722 | 4135154 | ||
BC015011.1 | - | 14 | 1714 | |||
BT007338.1 | - | 14 | 1716 | |||
D84224.1 | - | 14 | 1739 | |||
X94754.1 | - | 14 | 1723 | |||
Z49216.1 | - | 5 | 450 |