Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem167b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026198.2 | NP_080474.2 | 2 | 269 | ||
GenBank mRNA | AK008415.1 | - | 2 | 182 | ||
AK046697.1 | - | 2 | 170 | |||
AK155399.1 | - | 2 | 165 | |||
BC050932.1 | - | 2 | 173 | 6811263 | ||
BC055449.1 | - | 2 | 186 | 3993993 | ||
BQ560740.1 | - | 2 | 269 |