Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026200.3 | NP_080476.2 | 2 | 936 | ||
GenBank mRNA | AK012275.1 | - | 2 | 731 | ||
AK034091.1 | - | 2 | 829 | |||
AK038450.1 | - | 2 | 923 | |||
AK076120.1 | - | 2 | 705 | |||
AK139482.1 | - | 2 | 772 | |||
BC117819.1 | - | 2 | 825 | |||
BC128477.1 | - | 2 | 825 | 40092817 | ||
BC128478.1 | - | 2 | 825 | 40092823 |